Skip to Content
MilliporeSigma
All Photos(1)

Documents

388963

Sigma-Aldrich

Trihexyl O-acetylcitrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C[CO2(CH2)5CH3][CH2CO2(CH2)5CH3]2
CAS Number:
Molecular Weight:
486.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.447 (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

CCCCCCOC(=O)CC(CC(=O)OCCCCCC)(OC(C)=O)C(=O)OCCCCCC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyungjoo Kim et al.
Environmental research, 175, 316-322 (2019-05-31)
Phthalates are mainly used as binders and plasticizers in various industrial products including detergents, surfactants, waxes, paints, pharmaceuticals, food products, and cosmetics. However, they have been reported to be endocrine disruptors, which are chemicals that can mimic or disturb endocrines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service