Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

387797

Sigma-Aldrich

4-Bromo-N,N-bis(trimethylsilyl)aniline

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$119.00
25 ML
$449.00

$119.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$119.00
25 ML
$449.00

About This Item

Linear Formula:
BrC6H4N[Si(CH3)3]2
CAS Number:
Molecular Weight:
316.38
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$119.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

bp

156-158 °C/23 mmHg (lit.)

density

1.121 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

C[Si](C)(C)N(c1ccc(Br)cc1)[Si](C)(C)C

InChI

1S/C12H22BrNSi2/c1-15(2,3)14(16(4,5)6)12-9-7-11(13)8-10-12/h7-10H,1-6H3

InChI key

YJYMZGDBOMZWSR-UHFFFAOYSA-N

Application

4-Bromo-N,N-bis(trimethylsilyl)aniline is an N-protected aryl reagent used in the synthesis of:
  • Hydrophilic conjugated porphyrin dimers for one-photon and two-photon photodynamic therapy.[1]
  • Functionalized organometallic polymer, poly(ferrocenylsilane).[2]
  • Borylanilines such as 4-(dimesitylboryl)aniline and 4-(dimesitylboryl)-3,5-dimethylaniline.[3]
  • Silicon-containing oligomeric poly(imido-amides) (PIAs).[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Revisiting Borylanilines: Unique Solid-State Structures and Insight into Photophysical Properties.
Sudhakar P, et al.
Organometallics, 32(10), 3129-3133 (2013)
Synthesis of hydrophilic conjugated porphyrin dimers for one-photon and two-photon photodynamic therapy at NIR wavelengths.
Balaz M, et al.
Organic & Biomolecular Chemistry, 7(5), 874-888 (2009)
Silicon-containing oligomeric poly (imido-amides) with amino moieties. Synthesis, characterization and thermal studies.
Tagle L H, et al.
Royal Society of Chemistry Advances, 4(57), 30197-30210 (2014)
Versatile and convenient routes to functionalized poly (ferrocenylsilanes).
Jakle F, et al.
Macromolecular Rapid Communications, 21(18), 1291-1296 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service