Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

387576

Sigma-Aldrich

Decanoyl chloride

technical grade, 90%

Synonym(s):

Capric acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COCl
CAS Number:
Molecular Weight:
190.71
Beilstein:
507055
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

technical grade

vapor density

6.57 (vs air)

Assay

90%

refractive index

n20/D 1.441 (lit.)

bp

94-96 °C/5 mmHg (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC(Cl)=O

InChI

1S/C10H19ClO/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3

InChI key

IPIVAXLHTVNRBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service