Skip to Content
MilliporeSigma
All Photos(1)

Documents

386618

Sigma-Aldrich

Copper(II) 1,4,8,11,15,18,22,25-octabutoxy-29H,31H-phthalocyanine

Dye content 95 %

Synonym(s):

1,4,8,11,15,18,22,25-Octabutoxyphthalocyanine copper(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H80CuN8O8
CAS Number:
Molecular Weight:
1152.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, 95%

mp

184-187 °C (lit.)

λmax

740 nm

SMILES string

CCCCOc1ccc(OCCCC)c2c3nc(nc4n5[Cu]n6c(n3)c7c(OCCCC)ccc(OCCCC)c7c6nc8nc(nc5c9c(OCCCC)ccc(OCCCC)c49)c%10c(OCCCC)ccc(OCCCC)c8%10)c12

InChI

1S/C64H80N8O8.Cu/c1-9-17-33-73-41-25-26-42(74-34-18-10-2)50-49(41)57-65-58(50)70-60-53-45(77-37-21-13-5)29-30-46(78-38-22-14-6)54(53)62(67-60)72-64-56-48(80-40-24-16-8)32-31-47(79-39-23-15-7)55(56)63(68-64)71-61-52-44(76-36-20-12-4)28-27-43(75-35-19-11-3)51(52)59(66-61)69-57;/h25-32H,9-24,33-40H2,1-8H3;/q-2;+2

InChI key

GUXRYGQKVMJNDV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naveen Rawat et al.
Scientific reports, 5, 16536-16536 (2015-11-13)
The origins of spin exchange in crystalline thin films of Copper Octabutoxy Phthalocyanine (Cu-OBPc) are investigated using Magnetic Circular Dichroism (MCD) spectroscopy. These studies are made possible by a solution deposition technique which produces highly ordered films with macroscopic grain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service