Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

385352

Sigma-Aldrich

1,4-Difluoro-2,5-dimethoxybenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H2(OCH3)2
CAS Number:
Molecular Weight:
174.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

123-125 °C (lit.)

functional group

fluoro

SMILES string

COc1cc(F)c(OC)cc1F

InChI

1S/C8H8F2O2/c1-11-7-3-6(10)8(12-2)4-5(7)9/h3-4H,1-2H3

InChI key

QNQUBUBFPGHXAL-UHFFFAOYSA-N

General description

1,4-Difluoro-2,5-dimethoxybenzene is an alkoxy arene. It undergoes 1:3 reaction with TaF5 in chloroform at 40-50°C to afford long-lived radical cation salts [1,4-F2-2,5-(MeO)2C6H2][Ta2F11].[1]

Application

1,4-Difluoro-2,5-dimethoxybenzene is suitable reagent used in a study to investigate the direct anodic electrodeposition of polypyrrole, poly(3,4-ethylenedioxythiophene) and polythiophene on Cu electrodes by employing a corrosion inhibitor.[2] It may be used in the following studies:
  • Synthesis of 7-fluoro-5,8-dimethoxy-1-naphthol.[3]
  • As novel precursor for the iterative double benzyne-furan Diels-Alder reactions.[4]
  • Synthesis of 1,4-bis(phosphino)-2,5-difluoro-3,6-dimethoxybenzenes.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gillian E Morton et al.
The Journal of organic chemistry, 70(9), 3525-3529 (2005-04-23)
[reaction: see text] The use of 1,4-difluoro-2,5-dimethoxybenzene as a novel precursor for iterative two-directional benzyne-furan Diels-Alder reactions, using a range of 2- and 3-substituted furans, is reported. Substituted oxabenzonorbornadienes were synthesized following the initial Diels-Alder reaction, which upon ring opening
Electrodeposition of Conducting Polymers on Copper in Nonaque-ous Media by Corrosion Inhibition.
Lee S and Lee H.
Journal of Electrochemical Science and Technology, 3(2), 85-89 (2012)
Fabio Marchetti et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(41), 13962-13969 (2013-08-21)
The 1:3 reactions of the alkoxy arenes 1,4-(MeO)2 C6 H4 and 1,4-F2 -2,5-(MeO)2 C6 H2 with TaF5 in chloroform at 40-50 °C resulted in formation in about 35 % yield of the long-lived radical cation salts [1,4-(MeO)2 C6 H4 ][Ta2 F11 ]
Gillian E Morton et al.
Organic letters, 8(13), 2859-2861 (2006-06-16)
[reaction: see text] 7-Fluoro-5,8-dimethoxy-1-naphthol, prepared from the lithiation and benzyne formation from 1,4-difluoro-2,5-dimethoxybenzene and Diels-Alder cycloaddition with furan, was sequentially C-glycosidated under Suzuki conditions and O-glycosidated using di-O-acetyl-L-rhamnal to provide the corresponding beta-naphthyl C,O-disaccharide. Further lithiation, benzyne formation, and cycloaddition
1, 4-Bis (phosphine)-2, 5-difluoro-3, 6-dihydroxybenzenes and their P-oxides: Syntheses, structures, ligating and electronic properties.
Pignotti LR, et al.
Journal of Organometallic Chemistry, 693(20), 3263-3272 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service