Skip to Content
MilliporeSigma
All Photos(1)

Documents

384372

Sigma-Aldrich

1,7-Dichloro-octamethyltetrasiloxane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl[Si(CH3)2O]3Si(CH3)2Cl
CAS Number:
Molecular Weight:
351.52
Beilstein:
1784800
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.405 (lit.)

bp

222 °C (lit.)

mp

−62 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(Cl)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)Cl

InChI

1S/C8H24Cl2O3Si4/c1-14(2,9)11-16(5,6)13-17(7,8)12-15(3,4)10/h1-8H3

InChI key

UHRAUGIQJXURFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemistry Letters (Jpn), 2133-2133 (1990)
Yun-Seok Choi et al.
Nature methods, 16(8), 771-777 (2019-07-17)
Ubiquitin (Ub) conjugation is an essential post-translational modification that affects nearly all proteins in eukaryotes. The functions and mechanisms of ubiquitination are areas of extensive study, and yet the dynamics and regulation of even free (that is, unconjugated) Ub are
Naciye Esma Tirtom et al.
Physical chemistry chemical physics : PCCP, 22(21), 11928-11935 (2020-05-21)
Polyamines are naturally occurring cationic molecules in cells. In addition to their roles in modulating gene expression and cell proliferation, they have been shown to stimulate DNA recombination. The molecular mechanism for stimulation is not clear. We utilized single-molecule tethered
Cathy J Anderson et al.
Cell reports, 26(7), 1681-1690 (2019-02-14)
Histone H3 lysine 79 (H3K79) methylation is enriched on actively transcribed genes, and its misregulation is a hallmark of leukemia. Methylation of H3K79, which resides on the structured disk face of the nucleosome, is mediated by the Dot1L methyltransferase. Dot1L

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service