Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

384356

Sigma-Aldrich

Ethyl 4-hydroxy-7-trifluoromethyl-3-quinolinecarboxylate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10F3NO3
CAS Number:
Molecular Weight:
285.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

>300 °C (lit.)

SMILES string

CCOC(=O)c1cnc2cc(ccc2c1O)C(F)(F)F

InChI

1S/C13H10F3NO3/c1-2-20-12(19)9-6-17-10-5-7(13(14,15)16)3-4-8(10)11(9)18/h3-6H,2H2,1H3,(H,17,18)

InChI key

YHUWNMJUAZJACG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 4-hydroxy-7-trifluoromethyl-3-quinolinecarboxylate, a heterocyclic building block, is a quinoline derivative.These derivatives have antiseptic, antipyretic and antiperiodic action and possess various pharmacological applications.

Application

Ethyl 4-hydroxy-7-trifluoromethyl-3-quinolinecarboxylate may be used in the preparation of novel rare earth (Eu 3+, Tb3+, Gd3+, Sm3+ and Dy3+) complexes.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photophysical properties of novel rare earth complexes with ethyl 4 hydroxy 7 trifluoromethyl 3 quinolinecarboxylate.
Honjie, et al.
Chinese Journal of Inorganic Chemistry / Wu Ji Hua Xue Xue Bao, 4 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service