Skip to Content
MilliporeSigma
All Photos(1)

Documents

383643

Sigma-Aldrich

Ethyl 5-hydroxy-3-methyl-4-isoxazolecarboxylate sodium salt hydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8NNaO4 · xH2O
CAS Number:
Molecular Weight:
193.13 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

276-277 °C (lit.)

SMILES string

[Na+].[H]O[H].CCOC(=O)c1c(C)noc1[O-]

InChI

1S/C7H9NO4.Na.H2O/c1-3-11-6(9)5-4(2)8-12-7(5)10;;/h10H,3H2,1-2H3;;1H2/q;+1;/p-1

InChI key

FUQGYGTVHBHPSD-UHFFFAOYSA-M

General description

Ethyl 5-hydroxy-3-methyl-4-isoxazolecarboxylate sodium salt hydrate is a heterocyclic building block. Preparation of ethyl 5-hydroxy-3-methyl-4-isoxazolecarboxylate sodium salt from diethyl acetylmalonate has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient synthesis of N-monoalkylated hydroxylamines.
Doleschall G.
Tetrahedron Letters, 28(26), 2993-2994 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service