Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

382442

Sigma-Aldrich

Ethyl (R)-(−)-3-(2,2-dimethyl-1,3-dioxolan-4-yl)-trans-2-propenoate

97%

Synonym(s):

Ethyl (R)-trans-3-(2,2-dimethyl-1,3-dioxolan-4-yl)propenoate, Ethyl (R)-trans-4,5-O-isopropylidene-4,5-dihydroxy-2-pentenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O4
CAS Number:
Molecular Weight:
200.23
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]20/D −41°, c = 1 in chloroform

refractive index

n20/D 1.452 (lit.)

density

1.05 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)\C=C\[C@@H]1COC(C)(C)O1

InChI

1S/C10H16O4/c1-4-12-9(11)6-5-8-7-13-10(2,3)14-8/h5-6,8H,4,7H2,1-3H3/b6-5+/t8-/m1/s1

InChI key

GZVXALXOWVXZLH-HQZHTGGTSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service