Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

381799

Sigma-Aldrich

2-Bromo-3-nitrotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)Br
CAS Number:
Molecular Weight:
216.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

135-136 °C/8 mmHg (lit.)

mp

38-41 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1Br)[N+]([O-])=O

InChI

1S/C7H6BrNO2/c1-5-3-2-4-6(7(5)8)9(10)11/h2-4H,1H3

InChI key

GCAAVRIWNMTOKB-UHFFFAOYSA-N

General description

Crystal structure of 2-bromo-3-nitrotoluene has been studied and the dihedral angle between the nitro group and the phenyl ring is reported to be 54.1(4)°.

Application

2-Bromo-3-nitrotoluene may be used in the preparation of unsymmetrical dimethyl dinitro biphenyl, via Ullmann reaction. It may be used as starting material in the synthesis of 10-methylfluoranthene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Li et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 10), o2641-o2641 (2011-11-08)
In the title compound, C(7)H(6)BrNO(2), the dihedral angle between the nitro group and the phenyl ring is 14.9 (11)°.
675. Synthesis of fluoranthenes. Part IV. 10-and 11-Substituted fluoranthenes.
HorwoodaTucker S.
Journal of the Chemical Society, 3213-3216 (1949)
Ullmann Synthesis of Six Dimethyldinitrobiphenyls and their Reduction to the Corresponding Diaminodimethylbiphenyls1.
Carlin RB and Foltz GE.
Journal of the American Chemical Society, 78(9), 1997-2000 (1956)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service