Skip to Content
MilliporeSigma
All Photos(1)

Documents

381381

Sigma-Aldrich

Allyl methyl carbonate

98%

Synonym(s):

Carbonic acid, methyl 2-propenyl ester, Methyl allyl carbonate, methyl prop-2-enyl carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2OCOOCH3
CAS Number:
Molecular Weight:
116.12
Beilstein:
1746960
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

refractive index

n20/D 1.406 (lit.)

bp

59-60 °C/35 mmHg (lit.)

density

1.022 g/mL at 25 °C (lit.)

SMILES string

COC(=O)OCC=C

InChI

1S/C5H8O3/c1-3-4-8-5(6)7-2/h3H,1,4H2,2H3

InChI key

YHLVIDQQTOMBGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the Pd(0)-mediated synthesis of allyl aryl ethers and sulfides, and α,β-unsaturated carbonyls.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synlett, 725-725 (1992)
Tetrahedron, 50, 10321-10321 (1994)
Tetrahedron, 42, 2971-2971 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service