Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

381322

Sigma-Aldrich

4,4′-Difluorobenzil

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COCOC6H4F
CAS Number:
Molecular Weight:
246.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

120-122 °C (lit.)

SMILES string

Fc1ccc(cc1)C(=O)C(=O)c2ccc(F)cc2

InChI

1S/C14H8F2O2/c15-11-5-1-9(2-6-11)13(17)14(18)10-3-7-12(16)8-4-10/h1-8H

InChI key

BRKULQOUSCHDGS-UHFFFAOYSA-N

Gene Information

General description

4,4′-Difluorobenzil is a benzil derivative. Its reaction with benzylurea and phenethylurea has been investigated. It is reported to undergo condensation with Fe2(SH)2(CO)6 to afford the adduct Fe2[S2C2(OH)2(C6H4-4-F)2](CO)6.

Application

4,4′-Difluorobenzil is suitable for use in the preparation of 2,3-bis(4-fluorophenyl)thieno[3,4-b]pyrazine. It may be used in the synthesis of following:
  • poly(ether-α-diketone)s, via nucleophilic substitution reaction with bisphenol A
  • 4,4′-difluoro-2,3-diphenylpyrazine (DPPF) and 4,4′-difluoro-5-methyl 2,3-diphenylpyrazine (MDPPF), via direct condensation with ethylenediamine and 1,2-diaminopropane, respectively
  • 2,3-bis(4-fluorophenyl)quinoxaline, via direct condensation with 1,2-phenylenediamine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jane L Stanley et al.
Organometallics, 26(8), 1907-1911 (2008-07-02)
Reaction of Fe(2)(SH)(2)(CO)(6) and HCHO, which gives Fe(2)[(SCH(2))(2)NH](CO)(6) in the presence of NH(3), affords the possible intermediate Fe(2)(SCH(2)OH)(2)(CO)(6), which has been characterized crystallographically as its axial-equatorial isomer. Fe(2)(SCH(2)OH)(2)(CO)(6) was shown to react with ammonia and amines to give Fe(2)[(SCH(2))(2)NR](CO)(6) (R
Giulio G Muccioli et al.
Organic letters, 5(20), 3599-3602 (2003-09-26)
[reaction: see text] A new access to benzhydryl-phenylureas is described. These new interesting urea derivatives were obtained by reaction of substituted benzils with substituted phenylureas under microwave irradiation. Phenylthiourea, when reacted with benzil, gave 3-phenyl-thiohydantoin. Moreover, benzylurea, as phenethylurea, gave
Synthesis and Characterization of N-(2-Ethylhexyl) carbazole-2, 3-Bis (4-fluorophenyl)) thieno [3, 4-b] pyrazine Copolymer.
Li JC, et al.
Bull. Korean Chem. Soc., 31(7), 2073-2076 (2010)
Yellow organic light-emitting diodes based on phosphorescent iridium (III) pyrazine complexes: Fine tuning of emission color.
Ge G, et al.
Inorgorganica Chimica Acta, 362(7), 2231-2226 (2009)
Fu-Ming Hwang et al.
Inorganic chemistry, 44(5), 1344-1353 (2005-03-01)
Rational design and syntheses of four iridium complexes (1-4) bearing two substituted quinoxalines and an additional 5-(2-pyridyl) pyrazolate or triazolate as the third coordinating ligand are reported. Single-crystal X-ray diffraction studies of 1 reveal a distorted octahedral geometry, in which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service