Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

379484

Sigma-Aldrich

Sodium benzyloxide solution

1.0 M in benzyl alcohol

Synonym(s):

Sodium benzenemethanolate, Sodium benzylate, Sodium phenylmethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2ONa
CAS Number:
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
liquid
To inquire about this product 379484, please contact your local MilliporeSigma office or dealer. Contact Technical Service

grade

for analytical purposes

form

liquid

concentration

1.0 M in benzyl alcohol

bp

203-205 °C

density

1.077 g/mL at 25 °C

SMILES string

[Na+].[O-]Cc1ccccc1

InChI

1S/C7H7O.Na/c8-6-7-4-2-1-3-5-7;/h1-5H,6H2;/q-1;+1

InChI key

LFQULJPVXNYWAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service