Skip to Content
MilliporeSigma
All Photos(1)

Documents

378178

Sigma-Aldrich

4-Chlorobutyl chloroformate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2(CH2)4Cl
CAS Number:
Molecular Weight:
171.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.453 (lit.)

bp

89-90 °C/10 mmHg (lit.)

density

1.252 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCCCCOC(Cl)=O

InChI

1S/C5H8Cl2O2/c6-3-1-2-4-9-5(7)8/h1-4H2

InChI key

OTDWRXBRYNVCDN-UHFFFAOYSA-N

General description

4-Chlorobutyl chloroformate is an acid halide. Its enthalpy of vaporization at boiling point has been reported.
Generation of 4-chlorobutyl chloroformate is reported during the condensation of hydroxy acid with diphosgene.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 514-514 (2014)
Shi-Kai Tian et al.
Accounts of chemical research, 37(8), 621-631 (2004-08-18)
Insights into the role played by modified cinchona alkaloids in the Sharpless asymmetric dihydroxylation inspired studies of modified cinchona alkaloids as chiral organic catalysts that lead to the development of highly enantioselective alcoholyses for the desymmetrization, kinetic resolution, and dynamic
Malcolm J D'Souza et al.
International journal of molecular sciences, 21(12) (2020-06-25)
A previous study of the effect of a 2-chloro substituent on the rates and the mechanisms of the solvolysis of ethyl chloroformate is extended to the effect of a 3-chloro substituent on the previously studied solvolysis of propyl chloroformate and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service