Skip to Content
MilliporeSigma
All Photos(1)

Documents

377325

Sigma-Aldrich

N-[Bis(methylthio)methylene]glycine methyl ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3S)2C=NCH2CO2CH3
CAS Number:
Molecular Weight:
193.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.547 (lit.)

density

1.217 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)CN=C(SC)SC

InChI

1S/C6H11NO2S2/c1-9-5(8)4-7-6(10-2)11-3/h4H2,1-3H3

InChI key

XZAPFOKZCXRSDH-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service