Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

377147

Sigma-Aldrich

1,1-Bis(diphenylphosphino)ethylene

98%

Synonym(s):

VDPP, Vinylidenebis(diphenylphosphine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)2P]2C=CH2
CAS Number:
Molecular Weight:
396.40
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

98%

form

solid

reaction suitability

reagent type: ligand

mp

110-111 °C (lit.)

functional group

phosphine

SMILES string

C=C(P(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C26H22P2/c1-22(27(23-14-6-2-7-15-23)24-16-8-3-9-17-24)28(25-18-10-4-11-19-25)26-20-12-5-13-21-26/h2-21H,1H2

InChI key

GEGLBMPXRFOXTK-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Cyclometallated palladium diphosphane catalysts by Michael addition reactions
  • A-frame structures with palladacycles (chelating agent)
  • Functionalized bis-phosphine monoxide ligands catalyzed by chiral palladacycles
  • Tetranuclear palladacycles with bridging diphosphine ligands
  • Tetranuclear bromide-bridged Cu(I) complexes

  • Used as carbon-bridged diphosphine ligand in Cr-catalyzed ethylene tri- and tetramerization reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service