Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

376892

Sigma-Aldrich

2,3-Dimethylanthraquinone

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$101.00

$101.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$101.00

About This Item

Empirical Formula (Hill Notation):
C16H12O2
CAS Number:
Molecular Weight:
236.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$101.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

210-212 °C (lit.)

functional group

ketone

SMILES string

Cc1cc2C(=O)c3ccccc3C(=O)c2cc1C

InChI

1S/C16H12O2/c1-9-7-13-14(8-10(9)2)16(18)12-6-4-3-5-11(12)15(13)17/h3-8H,1-2H3

InChI key

KIJPZYXCIHZVGP-UHFFFAOYSA-N

General description

2,3-Dimethylanthraquinone is 2,3-dimethyl substituted anthraquinone and its synthesis has been reported.[1] Electron transfer rate constants for the reaction between radical anions of 2,3-dimethylanthraquinone with benzyl bromide has been evaluated.[2] The photoreactivity of 2,3-dimethylanthraquinone has been evaluated to investigate its usefulness as a photo-reagent for the analysis of ginsenosides using photoreduction fluorescence (PRF) detection method.[3]

Application

2,3-Dimethylanthraquinone may be used as starting reagent in the regioselective synthesis of 6-amino-2,3-anthracenedimethanol.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 3-Dimethylanthraquinone.
Allen CFH and Alan Bell.
Organic Syntheses, 37-37 (1942)
Regioselective syntheses of 5-and 6-aminoanthracene-2, 3-dimethanol.
Sun S and Desper J.
Tetrahedron, 54(3), 411-422 (1998)
Is samarium diiodide an inner-or outer-sphere electron donating agent?
Enem?rke RJ, et al.
Chemical Communications (Cambridge, England), 4, 343-344 (1999)
Photoreactivity of anthraquinones for the analysis of ginsenosides using photoreduction fluorescence detection-HPLC
Park MK, et al.
Archives of Pharmacal Research, 19(6), 562-565 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service