Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

376582

Sigma-Aldrich

3-Chloro-4-fluorobenzonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)CN
CAS Number:
Molecular Weight:
155.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

69-71 °C (lit.)

functional group

chloro
fluoro
nitrile

SMILES string

Fc1ccc(cc1Cl)C#N

InChI

1S/C7H3ClFN/c8-6-3-5(4-10)1-2-7(6)9/h1-3H

InChI key

VAHXXQJJZKBZDX-UHFFFAOYSA-N

General description

The FTIR and Raman spectra of 3-chloro-4-fluorobenzonitrile by ab initio HF and density functional method has been reported. Reaction of 3-chloro-4-fluorobenzonitrile with potassium fluoride in 1,3-dimethylimidazolidine-2-one (DMI) has been reported.

Application

3-Chloro-4-fluorobenzonitrile may be used in the preparation of ethyl 5-(2-chloro-4-cyanophenoxy)indoline-1-carboxylate and 3-chloro-4-(indolin-5-yloxy)benzonitrile.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Igarashi et al.
Chemical & pharmaceutical bulletin, 48(11), 1689-1697 (2000-11-22)
While searching for novel nonsteroidal inhibitors of human and rat prostatic 5alpha-reductases, we found a new series of indoline and aniline derivatives that showed potent inhibitory activities for both enzymes. Among them, 3-chloro-4-¿[1-(4-phenoxybenzyl)indolin-5-yl]oxylbenzoic acid (2e, YM-36117) showed a more potent
N Sundaraganesan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(3), 1134-1139 (2008-05-02)
In this work, the experimental and theoretical spectra of 3-chloro-4-fluoro benzonitrile (3C4FBN) were studied. The Fourier transform infrared and Fourier transform Raman spectra of 3C4FBN were recorded in the solid phase. The optimized geometry was calculated by HF and B3LYP
Synthesis of 3, 4-difluorobenzonitrile and monofluorobenzonitriles by means of halogen-exchange fluorination.
Suzuki H and Kimura Y.
Journal of Fluorine Chemistry, 52(3), 341-351 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service