Skip to Content
MilliporeSigma
All Photos(1)

Documents

376477

Sigma-Aldrich

4-Amino-5-hydroxy-2,7-naphthalenedisulfonic acid disodium salt

65%, technical grade

Synonym(s):

8-Amino-1-naphthol-3,6-disulfonic acid disodium salt, Disodium 4-amino-5-hydroxy-2,7-naphthalenedisulfonate, H Acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H4(OH)(SO3Na)2
CAS Number:
Molecular Weight:
363.27
Beilstein:
3863707
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

technical grade

concentration

65%

impurities

10-20% Na2SO4

SMILES string

[Na+].[Na+].Nc1cc(cc2cc(cc(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service