Skip to Content
MilliporeSigma
All Photos(1)

Documents

376442

Sigma-Aldrich

1,5-Hexadiene-3,4-diol, mixture of (±) and meso

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH(OH)CH(OH)CH=CH2
CAS Number:
Molecular Weight:
114.14
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.479 (lit.)

bp

125 °C/45 mmHg (lit.)

mp

14-16 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(C=C)C(O)C=C

InChI

1S/C6H10O2/c1-3-5(7)6(8)4-2/h3-8H,1-2H2

InChI key

KUQWZSZYIQGTHT-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service