Skip to Content
MilliporeSigma
All Photos(1)

Documents

374555

Sigma-Aldrich

2,4,6-Tris(dimethylaminomethyl)phenol

technical grade, 70%

Synonym(s):

DMP-30

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2NCH2]3C6H2OH
CAS Number:
Molecular Weight:
265.39
Beilstein:
795751
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 21 °C)

Assay

70%

impurities

≤30% 2,6-bis(dimethylaminomethyl)phenol

refractive index

n20/D 1.516 (lit.)

bp

130-135 °C/1 mmHg (lit.)

density

0.969 g/mL at 25 °C (lit.)

SMILES string

CN(C)Cc1cc(CN(C)C)c(O)c(CN(C)C)c1

InChI

1S/C15H27N3O/c1-16(2)9-12-7-13(10-17(3)4)15(19)14(8-12)11-18(5)6/h7-8,19H,9-11H2,1-6H3

InChI key

AHDSRXYHVZECER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service