Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

374520

Sigma-Aldrich

Diethyl (trichloromethyl)phosphonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2O)2P(O)CCl3
CAS Number:
Molecular Weight:
255.46
Beilstein:
1210640
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.463 (lit.)

bp

130-131 °C/14 mmHg (lit.)

density

1.362 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)C(Cl)(Cl)Cl

InChI

1S/C5H10Cl3O3P/c1-3-10-12(9,11-4-2)5(6,7)8/h3-4H2,1-2H3

InChI key

RVAQSYWDOSHWGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Addition reaction of diethyl (trichloromethyl)phosphonate to olefins by non-chain catalytic reactions catalyzed by copper amine complexes has been reported. Irradiation of diethyl (trichloromethyl)phosphonate in MeCN is reported to afford corresponding monoesters and olefins, via photochemical type II elimination reaction.

Application

Diethyl (trichloromethyl)phosphonate may be used in the synthesis of chlorovinyl phosphonates via reaction with aldehydes and ketones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemical type I and II elimination reactions of dialkyl (trichloromethyl) phosphonates.
Suzuki N, et al.
Bulletin of the Chemical Society of Japan, 53(5), 1421-1424 (1980)
Addition of diethyl trichloromethylphosphonate to olefins catalysed by copper complexes.
Villemin D, et al.
Tetrahedron Letters, 35(21), 3537-3538 (1994)
A novel synthesis of phosphonates from diethyl (trichloromethyl) phosphonate.
Lowen GT and Almond MR.
The Journal of Organic Chemistry, 59(!6), 4548-4550 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service