Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

372692

Sigma-Aldrich

Chromium silicide

−230 mesh, ≥99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrSi2
CAS Number:
Molecular Weight:
108.17
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99% trace metals basis

particle size

−230 mesh

SMILES string

[Cr++].[Si-]#[Si-]

InChI

1S/Cr.Si2/c;1-2/q+2;-2

InChI key

HIQCZYFMLUNWOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service