Skip to Content
MilliporeSigma
All Photos(2)

Documents

372641

Sigma-Aldrich

Nickel phosphide

−100 mesh, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni2P
CAS Number:
Molecular Weight:
148.36
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

reaction suitability

reagent type: catalyst
core: nickel

particle size

−100 mesh

SMILES string

[Ni]P=[Ni]

InChI

1S/2Ni.P

InChI key

FBMUYWXYWIZLNE-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service