Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

372323

Sigma-Aldrich

(S)-(−)-α−Methyl-1-naphthalenemethanol

99%

Synonym(s):

(S)-(−)-1-(1-Naphthyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH(CH3)OH
CAS Number:
Molecular Weight:
172.22
Beilstein:
2045009
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

optical activity

[α]20/D −77°, c = 1 in methanol

mp

47-49 °C (lit.)

SMILES string

C[C@H](O)c1cccc2ccccc12

InChI

1S/C12H12O/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-9,13H,1H3/t9-/m0/s1

InChI key

CDRQOYRPWJULJN-VIFPVBQESA-N

Gene Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingo Bichlmaier et al.
Journal of medicinal chemistry, 49(5), 1818-1827 (2006-03-03)
A set of 28 enantiomers comprising rigid and flexible secondary alcohols was synthesized by the asymmetric Corey-Bakshi-Shibata reduction. The enantiomerically pure alcohols were subjected to enzymatic glucuronidation assays employing the human UDP-glucuronosyltransferases (UGTs) 2B7 and 2B17. Both UGTs displayed high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service