Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

372129

Sigma-Aldrich

2,4-Bis(α,α-dimethylbenzyl)phenol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5C(CH3)2]2C6H3OH
CAS Number:
Molecular Weight:
330.46
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

206 °C/15 mmHg (lit.)

mp

63-65 °C (lit.)

SMILES string

CC(C)(c1ccccc1)c2ccc(O)c(c2)C(C)(C)c3ccccc3

InChI

1S/C24H26O/c1-23(2,18-11-7-5-8-12-18)20-15-16-22(25)21(17-20)24(3,4)19-13-9-6-10-14-19/h5-17,25H,1-4H3

InChI key

FMUYQRFTLHAARI-UHFFFAOYSA-N

General description

2,4-Bis(α,α-dimethylbenzyl)phenol is reported to possess antihistaminic activity. Structure of 2,4-bis(α,α-dimethylbenzyl)phenol is reported to have a torsion angle of 129.95(13)° for the C-C bond that connects the benzyl carbon to the phenol ring ortho to the OH group.

Application

2,4-Bis(α,α-dimethylbenzyl)phenol may be used in the preparation of a novel ligand, 2,2′-methylenebis(4,6-di(1-methyl-1-phenylethyl)phenol) (MMPEP-H2).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan
Acta crystallographica. Section C, Crystal structure communications, 56 (Pt 8), 1046-1047 (2000-08-16)
The structure of the title compound, 2, 4-bis(1-methyl-1-phenylethyl)phenol, C(24)H(26)O, was found to have a torsion angle of 129.95 (13) degrees for the C-C bond that connects the benzyl carbon to the phenol ring ortho to the OH group. A value
Zhuo Wang et al.
Journal of chromatography. A, 1616, 460793-460793 (2019-12-24)
With 4,4'-diaminoterphenyl and phloroglucinol as the monomers, Azo-linked porous organic polymer (PP) was prepared by a green mild azo reaction. Azo-PP is rich in phenolic hydroxyl groups and benzene rings, which can form intermolecular hydrogen bonds with the phenolic hydroxyl
A highly efficient catalyst for the ?living? and ?immortal? polymerization of e-caprolactone and L-lactide.
Liu Y-C, et al.
Macromolecules, 34(18), 6196-6201 (2001)
María J Duart et al.
Journal of medicinal chemistry, 49(12), 3667-3673 (2006-06-09)
Molecular topology was used to achieve a mathematical model capable of classifying compounds according to their antihistaminic activity and low sedative effects. By application of this model of activity to databases containing chemical reagents and drugs exhibiting other pharmacological activity
Alan J Bergmann et al.
Analytical and bioanalytical chemistry, 412(19), 4527-4536 (2020-05-28)
Food contact materials (FCM) may contain complex mixtures of estrogenic chemicals. A yeast estrogen screen performed on high performance thin-layer chromatography plates (planar-YES, P-YES) is promising for analysis of such mixtures, as it could allow for better elucidation of effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service