Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

372056

Sigma-Aldrich

Oxazine 170 perchlorate

Dye content 95 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22ClN3O5
CAS Number:
Molecular Weight:
431.87
Beilstein:
5371613
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, 95%

mp

260 °C (dec.) (lit.)

λmax

624 nm

SMILES string

OCl(=O)(=O)=O.CCNc1cc2OC3=CC(=N/CC)\c4ccccc4C3=Nc2cc1C

InChI

1S/C21H21N3O.ClHO4/c1-4-22-16-11-19-18(10-13(16)3)24-21-15-9-7-6-8-14(15)17(23-5-2)12-20(21)25-19;2-1(3,4)5/h6-12,22H,4-5H2,1-3H3;(H,2,3,4,5)/b23-17+;

InChI key

YCGJPLISNURWCC-YFZNFVDXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxazine 170 perchlorate (O17) is an organic dye with high fluorescence efficiency. It is used as a laser dye with an absorption band at 621 nm.[1]

Application

O17 can be used as a dopant with poly(methyl methacrylate) (PMMA) which can be applied in fabrication of polymeric optical fibers.[1] It forms a composite membrane with ethyl cellulose which can be used in the development of sensing membranes for urea.[2][3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication and calibration of Oxazine-based optic fiber sensor for detection of ammonia in water.
Jalal AH, et al.
Applied Optics, 51(17), 3768-3775 (2012)
Development of a ratiometric fluorescent urea biosensor based on the urease immobilized onto the oxazine 170 perchlorate-ethyl cellulose membrane.
Duong HD and Rhee JI
Talanta, 134(2), 333-339 (2015)
Ratiometric fluorescent l-arginine and l-asparagine biosensors based on the oxazine 170 perchlorate-ethyl cellulose membrane.
An K, et al.
Engineering in Life Sciences, 17(8), 847-856 (2017)
Luminescent properties of Oxazine 170 Perchlorate doped PMMA fiber.
Miluski P.
Fibers, 5(2), 15-15 (2017)

Articles

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service