Skip to Content
MilliporeSigma
All Photos(1)

Documents

371785

Sigma-Aldrich

Dimethyl 5-hydroxyisophthalate

98%

Synonym(s):

Dimethyl 5-hydroxybenzene-1,3-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3-1,3-(CO2CH3)2
CAS Number:
Molecular Weight:
210.18
Beilstein:
2650120
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

162-164 °C (lit.)

SMILES string

COC(=O)c1cc(O)cc(c1)C(=O)OC

InChI

1S/C10H10O5/c1-14-9(12)6-3-7(10(13)15-2)5-8(11)4-6/h3-5,11H,1-2H3

InChI key

DOSDTCPDBPRFHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl 5-hydroxyisophthalate undergoes a base-catalyzed nucleophilic oxirane ring-opening addition reaction with allyl glycidyl ether, to afford 5-substituted derivatives of isophthalic acid. Influence of third component, i.e. a series of 1,ω-alkanediols on the copolymerization reaction of dimethyl 5-hydroxyisophthalate with poly(ethylene glycol) has been studied. Lipase-catalyzed condensation polymerization of dimethyl 5-hydroxyisophthalate with polyethylene glycol is reported.

Application

Novel building block derived from dimethyl 5-hydroxyisophthalate was used to prepare sterically crowded polyether dendrons, via O-allylation/Claisen rearrangement reactions. Dimethyl 5-hydroxyisophthalate may be used in the preparation of:
  • bis(phebox) derivative of the cyclophosphazene
  • 5-{2-[2-(2-methoxyethoxy)ethoxy]ethoxy}benzene-1,3-dicarboxylic acid dimethyl ester
  • fullerene derivative bearing a triethylene glycol-type polar head group

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajesh Kumar et al.
Molecular diversity, 6(3-4), 287-295 (2004-04-08)
The use of enzymes in synthetic applications has increased dramatically in the recent years and the field of polymer science is part of this trend. Synthesis of a variety of polymers using lipase catalyzed (Candida antarctica) polymerization reactions has led
Biocatalytic ?green? Synthesis of PEG-based aromatic polyesters: Optimization of the substrate and reaction conditions.
Kumar R, et al.
Green Chemistry, 6(10), 516-520 (2004)
Jeny Karabline et al.
Organic & biomolecular chemistry, 10(24), 4788-4794 (2012-05-19)
Up to third-generation sterically crowded polyether dendrons were prepared on a solid support, using a novel building block derived from dimethyl 5-hydroxyisophthalate via O-allylation/Claisen rearrangement key steps. These dendrons underwent smooth disassembly to monomers, when subjected to acidic solution. The
Esther Frederick et al.
Langmuir : the ACS journal of surfaces and colloids, 34(36), 10739-10747 (2018-08-16)
Physisorbed self-assembled monolayers (SAMs) have been suggested as potential models for three-dimensional (3D) crystallization. This work studies the effect of altering the chain length of 5-alkoxyisophthalic acid (C nISA) on self-assembled morphology in both two-dimensional (2D) and 3D to explore
Denis Nilov et al.
Tetrahedron letters, 55(36), 5078-5081 (2014-08-26)
In the course of development of novel capping ligands with variable steric factor, which will be used as an organic coating for metal oxide nanoparticles, a base-catalyzed nucleophilic oxirane ring-opening addition reaction between dimethyl 5-hydroxyisophthalate and allyl glycidyl ether was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service