Skip to Content
MilliporeSigma
All Photos(1)

Documents

371238

Labclear gas filter

Refillable, weight 400 g(volume)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.51

L × D

14 in. × 2 in.

weight

400 g(volume)

Looking for similar products? Visit Product Comparison Guide

General description

Removes water, oil, and other materials from inert carrier gases such as N2, He, or Ar. Filters contain a mixture of indicating Drierite and 13X molecular sieves. Refillable filter tested at 120 psi up to 100°F, disposable filter tested at 125 psi up to 100°F. Supplied with 1/4 in. compression fittings.

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD
Labclear is a trademark of Labclear

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service