Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

370657

Sigma-Aldrich

3′-(Trifluoromethoxy)acetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4COCH3
CAS Number:
Molecular Weight:
204.15
Beilstein:
1873998
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.452 (lit.)

density

1.285 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1cccc(OC(F)(F)F)c1

InChI

1S/C9H7F3O2/c1-6(13)7-3-2-4-8(5-7)14-9(10,11)12/h2-5H,1H3

InChI key

UYHTUQHYGKAYJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3′-(Trifluoromethoxy)acetophenone is a meta-substituted acetophenone. Ruthenium-catalyzed reaction of carbon-hydrogen bonds in 3′-(trifluoromethoxy)acetophenone (m-(trifluoromethoxy)acetophenone) with olefins has been examined.

Application

3′-(Trifluoromethoxy)acetophenone (m-(trifluoromethoxy)acetophenone) may be used in the preparation of 4-(3′-trifluoromethoxyphenyl)-thiazol-2-yl ammonium iodide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium-Catalyzed Addition of Carbon-Hydrogen Bonds in Aromatic Ketones to Olefins. The Effect of Various Substituents at the Aromatic Ring.
Sonoda M, et al.
Bulletin of the Chemical Society of Japan, 70(12), 3117-3128 (1997)
Michael J Gorczynski et al.
Chemistry & biology, 14(10), 1186-1197 (2007-10-27)
The two subunits of core binding factor (Runx1 and CBFbeta) play critical roles in hematopoiesis and are frequent targets of chromosomal translocations found in leukemia. The binding of the CBFbeta-smooth muscle myosin heavy chain (SMMHC) fusion protein to Runx1 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service