Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

370142

Sigma-Aldrich

(S)-(−)-2-Hydroxy-N-methylsuccinimide

97%

Synonym(s):

(S)-(−)-3-Hydroxy-1-methyl-2,5-pyrrolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
Beilstein:
4350458
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Quality Level

Assay

97%

form

solid

optical activity

[α]20/D −75°, c = 3.5 in ethanol

mp

82-87 °C (lit.)

SMILES string

CN1C(=O)C[C@H](O)C1=O

InChI

1S/C5H7NO3/c1-6-4(8)2-3(7)5(6)9/h3,7H,2H2,1H3/t3-/m0/s1

InChI key

NLKACWKSCREJEL-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(-)-2-Hydroxy-N-methylsuccinimide may be used as a chiral auxiliary to enhance the diastereomeric excess of the Diels-Alder adducts formed in asymmetric Diels-Alder reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Diels-Alder addition of cyclopentadiene to chiral naphthoquinones.
Brimble MA, et al.
Tetrahedron Asymmetry, 9(7), 1239-1255 (1998)
Novel Potassium-Channel Openers: Preparation and Pharmacological Evaluation of Racemic and Optically Active N-(6-Amino-3-pyridyl)-N'-bicycloalkyl-N''-cyanoguanidine Derivatives
Eda M, et al.
Journal of Medicinal Chemistry, 37(13), 1983-1990 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service