Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

369667

Supelco

(R)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride

98%

Synonym(s):

(R)-(−)-MTPA-Cl, Mosher’s acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(OCH3)(CF3)COCl
CAS Number:
Molecular Weight:
252.62
Beilstein:
3591563
MDL number:
UNSPSC Code:
12000000

Assay

98%

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.47 (lit.)

bp

213-214 °C (lit.)

density

1.35 g/mL at 25 °C (lit.)

SMILES string

CO[C@](C(Cl)=O)(c1ccccc1)C(F)(F)F

InChI

1S/C10H8ClF3O2/c1-16-9(8(11)15,10(12,13)14)7-5-3-2-4-6-7/h2-6H,1H3/t9-/m0/s1

InChI key

PAORVUMOXXAMPL-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

doi:10.1038/nprot.2007.354

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas R Hoye et al.
Nature protocols, 2(10), 2451-2458 (2007-10-20)
This protocol details the most commonly used nuclear magnetic resonance (NMR)-based method for deducing the configuration of otherwise unknown stereogenic, secondary carbinol (alcohol) centers (R1R2CHOH (or the analogous amines where OH is replaced by NH2)). This 'Mosher ester analysis' relies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service