Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

369039

Sigma-Aldrich

4,4′-Diphenyltetrathiafulvalene

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12S4
CAS Number:
Molecular Weight:
356.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

form

solid

mp

204-206 °C (lit.)

SMILES string

S1C=C(S\C1=C2\SC=C(S2)c3ccccc3)c4ccccc4

InChI

1S/C18H12S4/c1-3-7-13(8-4-1)15-11-19-17(21-15)18-20-12-16(22-18)14-9-5-2-6-10-14/h1-12H/b18-17-

InChI key

UXYULNILNJCKTO-ZCXUNETKSA-N

General description

4,4′-Diphenyltetrathiafulvalene (DPTTF) is a heterocyclic building block. Formation of binary-component self-assembled monolayer (SAM) of DPTTF and n-tetradecane molecules with periodic strip-like phase separation structures on a highly oriented pyrolytic graphite surface has been reported. DPTTF capped gold nanoparticles having 3 to 5nm diameter have been prepared. FT-SERS studies of DPTTF adsorbed on chemically deposited silver films and excited with near infrared radiation have been reported.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miao Zhao et al.
Nanotechnology, 20(42), 425301-425301 (2009-09-26)
We report the formation of a binary-component self-assembled monolayer (SAM) comprising 4,4'-diphenyltetrathiafulvalene (DP-TTF) and n-tetradecane (n-C(14)H(30)) molecules with periodic strip-like phase separation structures on a highly oriented pyrolytic graphite (HOPG) surface. Scanning tunneling microscopy (STM) imaging reveals that ordered DP-TTF
Resmi et al.
Journal of colloid and interface science, 217(2), 395-402 (1999-09-02)
4,4'-Diphenyltetrathiafulvalene (DPTTF) capped gold nanoparticles of 3- to 5-nm diameter have been prepared and their thermal stability has been investigated by variable temperature FT-IR spectroscopy. The DPTTF molecules at the cluster surface undergo charge-transfer complexation with C(60) to form Au/DPTTF.C(60).
SERS studies on tetrathiafulvalene, diphenyltetrathiafulene and octahydrodibenzotetrathiafulvalene.
Joy VT and. Srinivasan TKK.
Chemical Physics Letters, 328(1), 221-226 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service