Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

368229

Sigma-Aldrich

Benzyl-N,N-dimethyldithiocarbamate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCS2CH2C6H5
CAS Number:
Molecular Weight:
211.35
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

39-41 °C (lit.)

SMILES string

CN(C)C(=S)SCc1ccccc1

InChI

1S/C10H13NS2/c1-11(2)10(12)13-8-9-6-4-3-5-7-9/h3-7H,8H2,1-2H3

InChI key

HLZBIKHKXIJIHX-UHFFFAOYSA-N

General description

Herbicidal action of benzyl N, N-dialkyldithiocarbamates is reported.[1] Performance of benzyl-N,N-dimethyldithiocarbamate as reversible addition-fragmentation chain transfer (RAFT) controller during RAFT polymerization of styrene in supercritical carbon dioxide is reported.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mode of the herbicidal action of benzyl N, N-dialkyldithiocarbamates.
WAKAMORI S.
Agricultural and Biological Chemistry, 37(2), 307-312 (1973)
Effect of stabilizer concentration and controller structure and composition on polymerization rate and molecular weight development in RAFT polymerization of styrene in supercritical carbon dioxide.
Jaramillo-Soto G, et al.
Polymer, 50(21), 5024-5030 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service