Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

367117

Sigma-Aldrich

5,7-Dibromo-2-methyl-8-quinolinol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Br2NO
CAS Number:
Molecular Weight:
316.98
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

form

powder

mp

126-130 °C (lit.)

InChI

1S/C10H7Br2NO/c1-5-2-3-6-7(11)4-8(12)10(14)9(6)13-5/h2-4,14H,1H3

InChI key

BNACJQWJZKPAPV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jakub Wantulok et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-02)
A new approach to the synthesis of selected quinolinecarbaldehydes with carbonyl groups located at C5 and/or in C7 positions is presented in this paper in conjunction with spectroscopic characterization of the products. The classical Reimer-Tiemann, Vilsmeier-Haack and Duff aldehyde synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service