Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

367001

Sigma-Aldrich

4,4′,4′′-Trimethoxytrityl chloride

technical grade

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$38.00
5 G
$154.00

$38.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$38.00
5 G
$154.00

About This Item

Linear Formula:
(CH3OC6H4)3CCl
CAS Number:
Molecular Weight:
368.85
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$38.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

technical grade

form

powder

mp

146-148 °C (lit.)

functional group

chloro

SMILES string

COc1ccc(cc1)C(Cl)(c2ccc(OC)cc2)c3ccc(OC)cc3

InChI

1S/C22H21ClO3/c1-24-19-10-4-16(5-11-19)22(23,17-6-12-20(25-2)13-7-17)18-8-14-21(26-3)15-9-18/h4-15H,1-3H3

InChI key

OBFCMKPFILBCSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,4′,4″-Trimethoxytrityl chloride can be used:
  • In one of the key synthetic steps for the preparation of two phosphoramidite building blocks.[1]
  • To prepare protein tagging molecule (3-[tris(4-methoxyphenyl)methylsulfanyl]propanoic acid by reacting with thioacetic acid.[2]
  • To prepare C-2 amino trimethoxytrityl substituted nucleoside, which is a key intermediate for the synthesis of nucleoside based capping agent.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An effective method for the synthesis of the cap structure of eukaryotic messenger ribonucleic acids
Kamimura T, et al.
Tetrahedron Letters, 25(25), 2683-2686 (1984)
Novel reagents for terminal phosphorylation and thiophosphorylation of synthetic oligonucleotides
Guzaev AP and Manoharan M
Tetrahedron Letters, 42(29), 4769-4773 (2001)
Mass-tag enhanced immuno-laser desorption/ionization mass spectrometry for sensitive detection of intact protein antigens
Lorey M, et al.
Analytical Chemistry, 87(10), 5255-5262 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service