Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

366951

Sigma-Aldrich

4-(Trifluoromethoxy)phenylhydrazine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4NHNH2·HCl
CAS Number:
Molecular Weight:
228.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

230 °C (lit.)

functional group

fluoro
hydrazine

SMILES string

Cl.NNc1ccc(OC(F)(F)F)cc1

InChI

1S/C7H7F3N2O.ClH/c8-7(9,10)13-6-3-1-5(12-11)2-4-6;/h1-4,12H,11H2;1H

InChI key

KQXZVSQCMVKMBK-UHFFFAOYSA-N

General description

4-(Trifluoromethoxy)phenylhydrazine hydrochloride is an organic building block.

Application

4-(Trifluoromethoxy)phenylhydrazine hydrochloride may be used in the synthesis of pyrazole-containing bisphosphonate (N-BPs) esters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoyue Xiang et al.
Organic & biomolecular chemistry, 10(38), 7730-7738 (2012-08-21)
An efficient general synthetic approach has been developed to synthesize pyrazole-containing bisphosphonate (N-BPs) esters from chromenone derivatives via a sequential two-step reaction in one pot at ambient temperature in good to excellent yields. This protocol provides a new convenient method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service