Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

366366

Sigma-Aldrich

(S)-5-(Hydroxymethyl)-2-pyrrolidinone

97%

Synonym(s):

L-Pyroglutaminol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80670
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +30°, c = 2 in ethanol

mp

79-80 °C (lit.)

SMILES string

OC[C@@H]1CCC(=O)N1

InChI

1S/C5H9NO2/c7-3-4-1-2-5(8)6-4/h4,7H,1-3H2,(H,6,8)/t4-/m0/s1

InChI key

HOBJEFOCIRXQKH-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 57, 2158-2158 (1992)
Tetrahedron, 48, 2143-2143 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service