Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

366226

Sigma-Aldrich

Diethyl (phthalimidomethyl)phosphonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16NO5P
CAS Number:
Molecular Weight:
297.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

60-64 °C (lit.)

functional group

imide
phosphonate

SMILES string

CCOP(=O)(CN1C(=O)c2ccccc2C1=O)OCC

InChI

1S/C13H16NO5P/c1-3-18-20(17,19-4-2)9-14-12(15)10-7-5-6-8-11(10)13(14)16/h5-8H,3-4,9H2,1-2H3

InChI key

IUZMHUAHKBHJFY-UHFFFAOYSA-N

Application

Reacant for:
  • Horner-Wadsworth-Emmons reactions
  • Preparation of macrocyclic phosphorous acid analogs as inhibitors of HCV-NS3 protease
  • Synthesis of dendrimer-based pH-responsive MRI contrast agent
Wittig carbonylating reagent.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 23, 43-43 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service