Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

365793

Sigma-Aldrich

2-Chloro-5-(trifluoromethyl)benzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CF3)CN
CAS Number:
Molecular Weight:
205.56
Beilstein:
1912184
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

210-212 °C (lit.)

mp

38-42 °C (lit.)

density

1.481 g/mL at 25 °C (lit.)

functional group

chloro
fluoro
nitrile

SMILES string

FC(F)(F)c1ccc(Cl)c(c1)C#N

InChI

1S/C8H3ClF3N/c9-7-2-1-6(8(10,11)12)3-5(7)4-13/h1-3H

InChI key

LCISFYAQKHOWBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Synthesis of 2-chloro-5-trifluoromethyl-benzonitrile, via Sandmeyer reaction of 2-chloro-5-trifluoromethylaniline with copper cyanide/sodium cyanide, is reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scale-up and safety evaluation of a Sandmeyer reaction.
Nielsen MA, et al.
Organic Process Research & Development, 8(6), 1059-1064 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service