Skip to Content
MilliporeSigma
All Photos(1)

Documents

365629

Supelco

(S,S)-(+)-Tetrandrine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H42N2O6
CAS Number:
Molecular Weight:
622.75
Beilstein:
877811
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D +285°, c = 1 in chloroform

mp

219-222 °C (lit.)

SMILES string

COc1ccc2C[C@@H]3N(C)CCc4cc(OC)c(OC)c(Oc5cc6[C@H](Cc7ccc(Oc1c2)cc7)N(C)CCc6cc5OC)c34

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuguang Ye et al.
Malaria journal, 12, 117-117 (2013-04-04)
In vitro evidence indicates that tetrandrine (TT) can potentiate the action of chloroquine 40-fold against choloquine-resistant Plasmodium falciparum. The key question emanating from that study is "would tetrandine and chloroquine be highly effective in a live Aotus monkey model with
Lihong Liu et al.
Journal of pharmaceutical and biomedical analysis, 61, 252-255 (2011-12-27)
A simple and selective method was developed for the simultaneous determination of tetrandrine and fangchinoline in herbal medicine by HPLC with electrochemical detection (ECD) on a bare glassy carbon electrode. The HPLC separation and ECD conditions have been optimized. The
Xiao-hua Xu et al.
Journal of Zhejiang University. Science. B, 13(11), 867-874 (2012-11-06)
To evaluate the effects of tetrandrine citrate, a novel tetrandrine salt with high water solubility, on the growth of imatinib (IM)-resistant chronic myeloid leukemia (CML) in vitro and in vivo, and reveal action molecular mechanisms. Cell viability in vitro was
Guo-Ying Zuo et al.
Molecules (Basel, Switzerland), 16(12), 9819-9826 (2011-11-26)
The antibacterial activity of two bisbenzylisoquinoline alkaloids, tetrandrine (Tet) and demethyltetrandrine (d-Tet), alone and in combination with the antibiotics ampicillin (AMP), azithromycin (AZM), cefazolin (CFZ) and levofloxacin (LEV) against 10 clinical isolates of staphylococcal chromosomal cassette mec (SCCmec) III type
Xiaolin Li et al.
International journal of nanomedicine, 7, 5183-5190 (2012-10-12)
Paclitaxel (Ptx), one of the most widely used anticancer agents, has demonstrated extraordinary activities against a variety of solid tumors. However, the therapeutic response of Ptx is often associated with severe side effects caused by its nonspecific cytotoxic effects and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service