Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

365335

Sigma-Aldrich

Etioporphyrin I nickel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36N4Ni
CAS Number:
Molecular Weight:
535.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

λmax

392 nm

SMILES string

CCc1c(C)c2cc3c(CC)c(C)c4cc5nc(cc6c(CC)c(C)c(cc1n2)n6[Ni]n34)c(C)c5CC

InChI

1S/C32H36N4.Ni/c1-9-21-17(5)25-14-30-23(11-3)19(7)27(35-30)16-32-24(12-4)20(8)28(36-32)15-31-22(10-2)18(6)26(34-31)13-29(21)33-25;/h13-16H,9-12H2,1-8H3;/q-2;+2/b25-14-,26-13-,27-16-,28-15-,29-13-,30-14-,31-15-,32-16-;

InChI key

SPKLBIKKHWTZSI-UGIVYKKHSA-N

General description

Nickel can be removed from nickel etioporphyrin (Ni-EP) by using supercritical water (SCW) at a temperature of 500°C and a pressure of 50MPa.

Application

Ni-EP can be used to fabricate a field effect transistor (FET).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel removal from nickel etioporphyrin (Ni-EP) using supercritical water in the absence of catalyst
Mandal PC, et al.
Fuel Processing Technology, 104, 67-72 (2012)
Field-effect transistor fabricated with nickel (II) etioporphyrin-I micrometer-sized crystals
Xu ZX, et al.
Applied Physics Letters, 93(22), 223305-223305 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service