Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

364967

Sigma-Aldrich

2,4,6-Trichlorophenyl chlorothionoformate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H2Cl4OS
CAS Number:
Molecular Weight:
275.97
MDL number:
UNSPSC Code:
12352103

Assay

95%

refractive index

n20/D 1.605 (lit.)

bp

146-147 °C/12 mmHg (lit.)

density

1.56 g/mL at 25 °C (lit.)

SMILES string

ClC(=S)Oc1c(Cl)cc(Cl)cc1Cl

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service