Skip to Content
MilliporeSigma
All Photos(1)

Documents

364878

Sigma-Aldrich

1-Bromocarbonyl-1-methylethyl acetate

≥95.0%

Synonym(s):

α-Acetoxyisobutyryl bromide, 2-Acetoxy-2-methylpropionyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC(CH3)2COBr
CAS Number:
Molecular Weight:
209.04
Beilstein:
2432585
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

refractive index

n20/D 1.457 (lit.)

bp

75-77 °C/12 mmHg (lit.)

density

1.431 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)OC(C)(C)C(Br)=O

InChI

1S/C6H9BrO3/c1-4(8)10-6(2,3)5(7)9/h1-3H3

InChI key

OOKAXSHFTDPZHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromocarbonyl-1-methylethyl acetate (α-Acetoxyisobutyryl bromide) participates in the conversion of adenosine to trans-3′(2′)-bromo-2′(3′)-acetates, via glycosyl cleavage.

Application

1-Bromocarbonyl-1-methylethyl acetate (α-Acetoxyisobutyryl bromide) may be used:
  • in the chemoenzymatic synthesis of (1R,2S)-1,2-epoxy-1,2-dihydroacridine (acridine 1,2-oxide)
  • in the synthesis of protected 3′-deoxyadenosine and 3′-deoxyguanosine
  • as reagent for deoxygenation of vicinal diols, as well as in the preparation of 2′,3′-dideoxycytidine and other dideoxy and deoxynucleosides from the corresponding ribo series.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mild conversion of vicinal diols to alkenes. Efficient transformation of ribonucleosides into 2'-ene and 2t', 3'-dideoxynucleosides.
Robins MJ, et al.
Tetrahedron Letters, 25(4), 367-370 (1984)
Aldrichimica Acta, 23, 83-83 (1990)
Efficient synthesis of protected 3'-deoxyadenosine and 3'-deoxyguanosine from adenosine and guanosine.
Cui Z, et al.
Tetrahedron Letters, 42(4), 561-563 (2001)
Biphenyl dioxygenase-catalysed cis-dihydroxylation of tricyclic azaarenes: chemoenzymatic synthesis of arene oxide metabolites and furoquinoline alkaloids.
Boyd DR, et al.
Royal Society of Chemistry Advances, 3(27), 10944-10955 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service