Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

363847

Sigma-Aldrich

2-Bromo-4-fluorotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(Br)F
CAS Number:
Molecular Weight:
189.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.528 (lit.)

bp

170-172 °C (lit.)

density

1.526 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Cc1ccc(F)cc1Br

InChI

1S/C7H6BrF/c1-5-2-3-6(9)4-7(5)8/h2-4H,1H3

InChI key

SFGFOJPGCOYQJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromo-4-fluorotoluene may be used in the preparation of meta-fluoro analog, (tris(5-fluoro-2-methylphenyl)phosphane, F-TOTP).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Hitce et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(3), 792-799 (2006-10-18)
A novel catalytic system, based on a mixture of palladium acetate and tris(5-fluoro-2-methylphenyl)phosphane (F-TOTP), has been designed for the intramolecular C--H functionalization of alkane segments. Among other analogues of tris(2-methylphenyl)phosphane (P(o-tol)(3)), F-TOTP was shown to have the optimal metal-bonding properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service