Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

363812

Sigma-Aldrich

4-(Decyloxy)benzoic acid

98%

Synonym(s):

4-(n -Decyloxy)benzoic acid, p -(Decyloxy)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9OC6H4CO2H
CAS Number:
Molecular Weight:
278.39
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

form

liquid crystal

mp

94-143 °C (lit.)

transition temp

nematic phase to isotropic phase 117-143 °C
smectic phase to nematic phase 94-117 °C

SMILES string

CCCCCCCCCCOc1ccc(cc1)C(O)=O

InChI

1S/C17H26O3/c1-2-3-4-5-6-7-8-9-14-20-16-12-10-15(11-13-16)17(18)19/h10-13H,2-9,14H2,1H3,(H,18,19)

InChI key

NZNICZRIRMGOFG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Sivaranjini et al.
Scientific reports, 8(1), 8891-8891 (2018-06-13)
A simple and effective approach for vertical alignment of liquid crystals (LCs) over a functionalized transparent flexible substrate is described. Surface characterization of this commercially available plastic substrate through X-ray photoelectron spectroscopy (XPS) and attenuated total reflection infrared spectroscopy (ATR-IR)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service