Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

363723

Sigma-Aldrich

(Bicyclo[2.2.1]hepta-2,5-diene)dichlororuthenium(II) polymer

Synonym(s):

Dichloro(norbornadiene)ruthenium, NSC 71325

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300

reaction suitability

core: ruthenium
reagent type: catalyst

InChI

1S/C7H6Cl2.Ru/c8-6-4-1-2-5(3-4)7(6)9;/h1-3H2;/q;+2

InChI key

YEEAFQBZFSVTRQ-UHFFFAOYSA-N

Application

Catalyst for:
  • Direct arylation of arene C-H bonds by proton abstraction
  • Regio- and stereoselective hydrosilylation of isoprene

Catalyst for synthesis of:
  • Vinyl carbamates from carbon dioxide and alkynes
  • O-beta-oxoalkylcarbamates

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service