Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

363529

Sigma-Aldrich

(R)-(−)-Glycidyl benzyl ether

99%

Synonym(s):

(R)-(−)-2-(Benzyloxymethyl)oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
Beilstein:
3588399
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]20/D −5.4°, c = 5 in toluene

refractive index

n20/D 1.517 (lit.)

density

1.077 g/mL at 25 °C (lit.)

functional group

ether
phenyl

storage temp.

2-8°C

SMILES string

C(OCc1ccccc1)[C@H]2CO2

InChI

1S/C10H12O2/c1-2-4-9(5-3-1)6-11-7-10-8-12-10/h1-5,10H,6-8H2/t10-/m0/s1

InChI key

QNYBOILAKBSWFG-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the preparation of the lactone fragment of compactin and mevinolin. Chiron in the preparation of syn-1,3-polyols, dideoxynucleosides, and a spiroacetal cyanohydrin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 539-539 (1989)
Journal of the Chemical Society. Chemical Communications, 291-291 (1993)
E Abushanab et al.
Journal of medicinal chemistry, 32(1), 76-79 (1989-01-01)
1',2'-seco-2',3'-Dideoxycytidine (12), -guanosine (14), -adenosine (16), and -inosine (18) were prepared from (R)-benzylglycidol as potential anti-HIV agents. When compared to ddAdo in protecting ATH8 cells, they were found to be inactive.
The Journal of Organic Chemistry, 53, 4495-4495 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service