Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36340

Sigma-Aldrich

2,3-Dichloropropionic acid

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CHClCOOH
CAS Number:
Molecular Weight:
142.97
Beilstein:
1721427
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

mp

48-52 °C

functional group

carboxylic acid
chloro

SMILES string

OC(=O)C(Cl)CCl

InChI

1S/C3H4Cl2O2/c4-1-2(5)3(6)7/h2H,1H2,(H,6,7)

InChI key

GKFWNPPZHDYVLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service