Skip to Content
MilliporeSigma
All Photos(1)

Documents

363359

Sigma-Aldrich

2-Butene, mixture of cis and trans

≥99%

Synonym(s):

β-Butylene, Pseudobutylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH3
CAS Number:
Molecular Weight:
56.11
Beilstein:
1718755
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1.9 mmHg ( 0 °C)

Assay

≥99%

expl. lim.

9.3 %

bp

1 °C (lit.)

mp

−140 °C (lit.)

SMILES string

C\C=C\C

InChI

1S/C4H8/c1-3-4-2/h3-4H,1-2H3/b4-3+

InChI key

IAQRGUVFOMOMEM-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Butene is an alkene. All-silica zeolite RUB-41, containing 8- and 10-membered rings, is reported to be capable of separating trans-2-butene and cis-2-butene from 1-butene. Asymmetric gydrozirconation of cis- and trans-2-butene is reported. Stereospecific addition of methylene to trans-2-butene is reported to afford trans-1,2dimethylcyclopropane while similar addition to cis-2-butene affords cis-1,2dimethylcyclopropane.

Recommended products

Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended.

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-29.2 °F - closed cup

Flash Point(C)

-34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylene, CH2. Stereospecific Reaction with cis-and trans-2-Butene.
Woodworth RC and Skell PS.
Journal of the American Chemical Society, 81(13), 3383-3386 (1959)
Yonghui Zhang et al.
Journal of the American Chemical Society, 125(29), 8746-8747 (2003-07-17)
Hydrozirconation of cis-2-butene with Cp*ZrHCl[N(t-Bu)C(Me)N(Et)], generated in situ through hydrogenolysis of Cp*ZrCl(SiMe2Ph)[N(t-Bu)C(Me)N(Et)] (5), proceeds in high yield to produce a 1:2 mixture of the kinetically stable, diastereomeric sec-butyl complexes, 3a and 3b. Hydrozirconation of trans-2-butene under identical conditions provides a
Ollie M Gonzalez-James et al.
Journal of the American Chemical Society, 134(4), 1914-1917 (2012-01-11)
An unusual intramolecular kinetic isotope effect (KIE) in the reaction of dichloroketene with cis-2-butene does not fit with a simple asynchronous cycloaddition transition state, but it can be predicted from trajectory studies on a bifurcating energy surface. The origin of
Robert R Arnts
Environmental science & technology, 42(20), 7663-7669 (2008-11-06)
The continuous addition of trans-2-butene to air containing ozone-reactive volatile and semivolatile organic compounds prior to sampling on Tenax-TA adsorbent was found to be an effective means of removing ozone and reducing analyte losses of ozone reactive biogenic volatile organic
Nadiya Bakhiya et al.
Archives of toxicology, 84(7), 563-578 (2010-03-20)
Furan is formed during commercial or domestic thermal treatment of food. The initial surveys of furan concentrations in heat-treated foods, published by European and US authorities, revealed the presence of relatively high furan levels in coffee, sauces, and soups. Importantly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service